

(-)-Isopinocampheylborane TMEDA complex ≥98.0% (by total nitrogen basis)
Catalog # TCIAI0797-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (with Total Nitrogen):min. 98.0 %
- Specific rotation [a]20/D:-55.0 to -62.0 deg (C=1、CHCl3)
Specification Test Results
About this item
:
:416.35 g/mol
UN:1325
ADR:4.1,II
:GHS02
:GHS07
CAS Number:67826-92-0
Formula:C₂₆H₅₄B₂N₂
MDL Number:MFCD00077801