
(R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) ≥98.0% (by GC)
Catalog # TCIAI0795-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:88.0 to 92.0 deg-C
- Specific rotation [a]20/D:+112.0 to +123.0 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:294.44 g/mol
:GHS07
CAS Number:131833-97-1
Formula:C₁₇H₃₀N₂O₂
MDL Number:MFCD07368371