Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-Iodophenyl trifluoromethanesulphonate ≥98.0% (by GC)
  TCIAI0770-5G
 :  TCI
 :  
 
2-Iodophenyl trifluoromethanesulphonate ≥98.0% (by GC)
  TCIAI0770-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :352.07 g/mol
 :GHS07
 :129112-26-1
 :C₇H₄F₃IO₃S
 :MFCD07784326