Imidazo[1,5-a]pyridine ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0753-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:52.0 to 57.0 deg-C
Specification Test Results
About this item
:
:118.14 g/mol
Storage Temperature:Freezer
CAS Number:274-47-5
Formula:C₇H₆N₂
MDL Number:MFCD03407375