(S)-(4-Isopropyloxazolin-2-yl)ferrocene ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0749-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Chelometric Titration):min. 98.0 %
- Specific rotation [a]20/D:-133.0 to -137.0 deg (C=1, EtOH)
Specification Test Results
About this item
:
:297.18 g/mol
CAS Number:162157-03-1
Formula:C₁₆H₁₉FeNO