Imidazolidinyl urea ≥90.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:39236-46-9
:
Formula:C₁₁H₁₆N₈O₈
MDL Number:MFCD00221482
:388.30 g/mol
Merck Index:14,04916
EINECS:254-372-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 90.0 %
Specification Test Results
Frequently Bought Together