
(Isocyanoimino)triphenylphosphorane ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:73789-56-7
:
Formula:C₁₉H₁₅N₂P
MDL Number:MFCD09038528
:302.32 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):8.80 to 9.70 %
Specification Test Results
Frequently Bought Together