
(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:131833-93-7
:
Formula:C₁₇H₃₀N₂O₂
MDL Number:MFCD00192243
:294.44 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:88.0 to 92.0 deg-C
- Specific rotation [a]20/D:-112 to -123 deg (C=1, CHCl3)
Specification Test Results
Frequently Bought Together