
Isoquinoline-3-carboxylic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:203626-75-9
Formula:C₁₀H₉NO₃
MDL Number:MFCD00075137
:191.183 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in hot Water:almost transparency
- Water:8.9 to 9.9 %
Specification Test Results
Frequently Bought Together