(+)-5,6-O-Isopropylidene-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15042-01-0
:
Formula:C₉H₁₂O₆
MDL Number:MFCD00010552
:216.19 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Iodometric Titration):min. 98.0 %
- Specific rotation [a]20/D:+8.0 to +12.0 deg (C=5, Methanol)
- Solubility in Methanol:almost transparency
Specification Test Results
Related Information
Frequently Bought Together