Dimethyl-4,5-imidazoledicarboxylate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3304-70-9
:
Formula:C₇H₈N₂O₄
MDL Number:MFCD00051338
:184.1500 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together