Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(1-Isopropoxyethyl)acetamide
N-(1-Isopropoxyethyl)acetamide ≥93.0% (by GC)
N-(1-Isopropoxyethyl)acetamide ≥93.0% (by GC)
N-(1-Isopropoxyethyl)acetamide ≥93.0% (by GC)
TCIAI0441-25G
:
TCI
:
N-(1-Isopropoxyethyl)acetamide ≥93.0% (by GC)
TCIAI0441-25G
:
TCI
:
Pk:
25 g
Seq:
3308
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 93.0 %
:
:
145.20 g/mol
:
89°C
:
115910-75-3
:
C₇H₁₅NO₂
:
MFCD00191526