Order Entry
Netherlands
ContactUsLinkComponent
Diallyl propyl isocyanurate ≥97.0% (by GC) stabilised
Irritant
Diallyl propyl isocyanurate ≥97.0% (by GC) stabilised
Catalog # TCIAI0409-5G
Supplier:  TCI
CAS Number:  
Irritant
Diallyl propyl isocyanurate ≥97.0% (by GC) stabilised
Catalog # TCIAI0409-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid to slightly cloudy liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :251.29 g/mol
Boiling Pt:136°C
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
 :GHS07
Storage Temperature:Refrigerator
CAS Number:5320-25-2
Formula:C₁₂H₁₇N₃O₃
MDL Number:MFCD00144514