Order Entry
Netherlands
ContactUsLinkComponent
p-Tolyl isovalerate ≥98.0% (by GC)
p-Tolyl isovalerate ≥98.0% (by GC)
Catalog # TCIAI0397-25ML
Supplier:  TCI
CAS Number:  
p-Tolyl isovalerate ≥98.0% (by GC)
Catalog # TCIAI0397-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :192.26 g/mol
Boiling Pt:125°C
EINECS:259-465-5
CAS Number:55066-56-3
Formula:C₁₂H₁₆O₂
MDL Number:MFCD00036648