Order Entry
Netherlands
ContactUsLinkComponent
cis-3-Hexenyl isovalerate ≥98.0% (by GC)
cis-3-Hexenyl isovalerate ≥98.0% (by GC)
Catalog # TCIAI0307-25ML
Supplier:  TCI
CAS Number:  
cis-3-Hexenyl isovalerate ≥98.0% (by GC)
Catalog # TCIAI0307-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :184.28 g/mol
Flash Pt:101°C
EINECS:252-404-3
CAS Number:35154-45-1
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00036533