
Diallyl isocyanurate ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0304-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Bromination Method):min. 98.0 %
- Melting point:145.0 to 150.0 deg-C
Specification Test Results
About this item
:
:209.21 g/mol
:GHS07
CAS Number:6294-79-7
:MFCD00059763
Formula:C₉H₁₁N₃O₃