
8-Hydroxyquinoline-2-carbaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14510-06-6
:
Formula:C₁₀H₇NO₂
MDL Number:MFCD00168962
:173.1681 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:96.0 to 100.0 deg-C
- Acid Value:max. 5.0
Specification Test Results
Frequently Bought Together