
3-Hydroxy-1-methacryloyloxyadamantane ≥99.0% (by GC), purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:115372-36-6
:
Formula:C₁₄H₂₀O₃
MDL Number:MFCD13152044
:236.31 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:89.0 to 93.0 deg-C
Specification Test Results
Frequently Bought Together