1,1'-(Hexane-1,6-diyl)bis[3-(2-nitrophenyl)urea] ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:2419249-92-4
:
Formula:C₂₀H₂₄N₆O₆
:444.45 g/mol
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together