

Ethyl isobutyrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-62-1
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00009165
:116.16 g/mol
Merck Index:13,03847
Boiling Pt:112…113 °C
Melting Pt:–88 °C
Density:0.86 g/cm³
Flash Pt:13 °C
Storage Temperature:Ambient
EINECS:202-595-4
UN:2385
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3860 to 1.3880 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together