
Thieno[3,2-b]pyridin-7-ol ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAH1603-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:233.0 to 237.0 deg-C
Specification Test Results
About this item
:
:151.183 g/mol
:GHS07
CAS Number:107818-20-2
Formula:C₇H₅NOS
MDL Number:MFCD01863636