
4',6'-Dimethoxy-2'-hydroxy-2(4'-methoxyphenyl)acetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:39604-68-7
:
Formula:C₁₇H₁₈O₅
MDL Number:MFCD03934798
:302.33 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:85.0 to 89.0 deg-C
Specification Test Results
Frequently Bought Together