
3-Hydroxy-4,4'-dimethyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:81998-07-4
:
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD28339195
:200.24 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:108.0 to 112.0 deg-C
Specification Test Results
Frequently Bought Together