
1,6-Hexylenediphosphonic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAH1536-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:203.0 to 208.0 deg-C
Specification Test Results
About this item
:
:246.14 g/mol
UN:3261
ADR:8,III
:GHS05
CAS Number:4721-22-6
Formula:C₆H₁₆O₆P₂
MDL Number:MFCD01630742