
7-Hydroxy-1-indanone ≥98.0% (by GC, titration analysis)
Catalog # TCIAH1526-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:110.0 to 114.0 deg-C
Specification Test Results
About this item
:
Boiling Pt:131°C
:148.161 g/mol
:GHS07
CAS Number:6968-35-0
Formula:C₉H₈O₂
MDL Number:MFCD01548387