Order Entry
Netherlands
ContactUsLinkComponent
Hydrocortisone 17-Butyrate ≥98.0% (by HPLC)
Irritant
Hydrocortisone 17-Butyrate ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
Irritant
Hydrocortisone 17-Butyrate ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    208.0 to 212.0 deg-C
  • Specific rotation [a]20/D:
    +48.0 to +52.0 deg (C=0.1,CHCl3) (calcd.on dried substance)
  • Drying loss:
    max. 1.0 %

Specification Test Results

Frequently Bought Together