
4-Hydroxy-2-methylphenylboronic acid (contains varying amounts of Anhydride)
: TCI
:493035-82-8
:
:C₇H₉BO₃
:MFCD03788424
:151.96 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 114.0 %
Frequently Bought Together