Order Entry
Netherlands
ContactUsLinkComponent
 
trans-2-Hexenyl valerate ≥94.0% (by GC)
Catalog # TCIAH1444-25ML
Supplier:  TCI
CAS Number:  
undefined
trans-2-Hexenyl valerate ≥94.0% (by GC)
Catalog # TCIAH1444-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 94.0 %

Specification Test Results

About this item

 :
 :184.28 g/mol
EINECS:260-439-0
CAS Number:56922-74-8
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00036547