
4-Cyanophenylhydrazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2863-98-1
:
Formula:C₇H₈ClN₃
MDL Number:MFCD00673994
:169.61 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Potassium iodate Method):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together