
2-Hydroxycyclohexanecarboxylic acid cis- and trans-mixture ≥98.0% (by GC, titration analysis)
: TCI
:609-69-8
:
:C₇H₁₂O₃
:MFCD02684437
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Neutralization titration):min. 98.0 %
- Melting point:67.0 to 71.0 deg-C
Frequently Bought Together