
4'-Hydroxyazobenzene-4-carboxylic acid hydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAH1400-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Water:max. 1.0 %
Specification Test Results
About this item
:
:242.23 g/mol
:GHS07
CAS Number:2497-38-3
Formula:C₁₃H₁₀N₂O₃
MDL Number:MFCD12198372