
4-(cis-4-Hydroxycyclohexyl)phenol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:370860-74-5
:
Formula:C₁₂H₁₆O₂
MDL Number:MFCD23380209
:192.26 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:207.0 to 211.0 deg-C
Specification Test Results
Frequently Bought Together