N-Hydroxy-3-[1-(phenylthio)methyl-1H-1,2,3-triazol-4-yl]benzamide ≥96.0% (by HPLC)
Catalog # TCIAH1340-5MG
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 96.0 area%
- Elemental analysis (Nitrogen):16.30 to 18.00 %
Specification Test Results
About this item
:
:326.37 g/mol
Storage Temperature:Refrigerator
CAS Number:1316652-41-1
Formula:C₁₆H₁₄N₄O₂S