4-(11-Hydroxyundecyloxy)benzaldehyde ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:124389-14-6
:
Formula:C₁₈H₂₈O₃
:292.42 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:62.0 to 66.0 deg-C
Specification Test Results
Frequently Bought Together