17beta-Hydroxy-17-methylandrosta-1,4-dien-3-one ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:72-63-9
:
Formula:C₂₀H₂₈O₂
MDL Number:MFCD00003650
:300.44 g/mol
Merck Index:14,05951
EINECS:200-787-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:162.0 to 166.0 deg-C
- Specific rotation [a]20/D:+8.0 to +10.0 deg (C=1, EtOH)
Specification Test Results
Frequently Bought Together