4-(3-Hydroxypropyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:205194-33-8
:
Formula:C₇H₁₅NO₃S
:193.26 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:70.0 to 74.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together