N-Hydroxynaphthalimide trifluoromethanesulphonate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:85342-62-7
:
Formula:C₁₃H₆F₃NO₅S
MDL Number:MFCD02683478
:345.25 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:211.0 to 215.0 deg-C
Specification Test Results
Frequently Bought Together