
1-Hexyl-3-methylimidazolium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAH1100-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
About this item
:
:316.34 g/mol
:GHS07
CAS Number:460345-16-8
Formula:C₁₁H₁₉F₃N₂O₃S
MDL Number:MFCD03427621