
4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13380-84-2
:
Formula:C₈H₁₄O₃
MDL Number:MFCD01734677
:158.20 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Purple powder to crystal
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together