
L-Histidine methyl ester dihydrochloride ≥98.0% (by HPLC, titration analysis)
: TCI
:7389-87-9
:
:C₇H₁₁N₃O₂
:MFCD00012701
:242.10 g/mol
:230-973-9
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:+8.0 to +11.0 deg (C=2, H2O)
- Solubility in Water:almost transparency
Frequently Bought Together