
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17419-81-7
:
Formula:C₇H₁₂O₃
MDL Number:MFCD00077974
:144.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min 98.0 % (total of isomer)
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together