7-Hydroxy-5-methyl-2-methylthio[1,2,4]triazolo[1,5-a]pyrimidine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:40775-78-8
:
Formula:C₇H₈N₄OS
MDL Number:MFCD00186154
:196.23 g/mol
EINECS:255-072-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Solubility in N,N-DMF:almost transparency
Specification Test Results
Frequently Bought Together