Order Entry
Netherlands
ContactUsLinkComponent
Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97.0% (by titrimetric analysis)
Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97.0% (by titrimetric analysis)
Catalog # TCIAH0917-25G
Supplier:  TCI
CAS Number:  
undefined
Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97.0% (by titrimetric analysis)
Catalog # TCIAH0917-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Chelometric Titration):
    min. 97.0 % (calcd.on anh.substance)
  • Solubility in Water:
    very faint turbidity
  • Water:
    max. 10.0 %

Specification Test Results

About this item

 :
 :274.33 g/mol (anhy)
CAS Number:135236-72-5
 :MFCD01318562
Formula:C₁₀H₁₈CaO₆·H₂O
MDL Number:MFCD01321171