3-Hydroxy-4-methyl-2(3H)-thiazolethione ≥98.0% (by titrimetric analysis) for source of alkyl radical
Catalog # TCIAH0902-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:94.0 to 97.0 deg-C
Specification Test Results
About this item
:
:147.21 g/mol
:GHS07
CAS Number:49762-08-5
Formula:C₄H₅NOS₂
MDL Number:MFCD00077793