Order Entry
Netherlands
ContactUsLinkComponent
3-Hepten-2-one ≥95.0% (by GC)
Flammable
3-Hepten-2-one ≥95.0% (by GC)
Catalog # TCIAH0838-25ML
Supplier:  TCI
CAS Number:  
Flammable
3-Hepten-2-one ≥95.0% (by GC)
Catalog # TCIAH0838-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :112.17 g/mol
Boiling Pt:156°C
Flash Pt:49°C
EINECS:214-278-8
UN:1224
ADR:3,III
 :GHS02
CAS Number:1119-44-4
Formula:C₇H₁₂O
MDL Number:MFCD00015564