
1,5-Hexadiene-3,4-diol ≥95.0% (by GC) stabilised
Catalog # TCIAH0807-10ML
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
Boiling Pt:198°C
Flash Pt:92°C
Storage Temperature:Refrigerator
EINECS:213-955-5
ADR:6.1,III
:Stabilised with hydroquinone (HQ)
:114.14 g/mol
UN:2810
:GHS06
CAS Number:1069-23-4
Formula:C₆H₁₀O₂
MDL Number:MFCD00081129