Order Entry
Netherlands
ContactUsLinkComponent
 
 
10-Hydroxybenzo[h]quinoline ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
 
10-Hydroxybenzo[h]quinoline ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    99.0 to 103.0 deg-C
  • Solubility in Acetone:
    almost transparency

 

Frequently Bought Together