
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53188-07-1
:
Formula:C₁₄H₁₈O₄
MDL Number:MFCD00006846
:250.29 g/mol
EINECS:258-422-8
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together