
2-Hydroxy-4-(methylthio)butyric acid 65 - 72% in water
Catalog # TCIAH0654-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Concentration (Neutralization titration):65.0 to 72.0 w/w%
Specification Test Results
About this item
:
:150.19 g/mol
Merck Index:14,05976
EINECS:209-523-0
:GHS05
CAS Number:583-91-5
Formula:C₅H₁₀O₃S
MDL Number:MFCD00070490