

Bis(hexafluoroacetylacetonato)cobalt(II) hydrate ≥95.0% (by titrimetric analysis)
Catalog # TCIAH0553-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Chelometric Titration):min. 95.0 % (calcd.on anh.substance)
- Water:max. 12.0 %
Specification Test Results
About this item
:
:473.04 g/mol
:GHS07
:GHS08
CAS Number:19648-83-0
Formula:C₁₀H₂CoF₁₂O₄
MDL Number:MFCD00042510